missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DDR2, GST Tag Recombinant Protein
DDR2 is a member of a subclass of RTKs and contains a distinct extracellular region encompassing a factor VIII-like domain.
Marca: Invitrogen™ PV4188
Detalles adicionales : Peso : 0.00900kg
Descripción
DDR1 and DDR2 have been shown to be potently inhibited by Gleevec.
Especificaciones
NP_006173, S642A | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
DDR2, TK, NTRKR3 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
79.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
DDR Kinase Family | |
Recombinant | |
Baculovirus |