missing translation for 'onlineSavingsMsg'
Learn More

DCK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17758-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16807724

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 23-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
Especificaciones

Especificaciones

DCK
Polyclonal
Unconjugated
DCK
DCK, deoxycytidine kinase
Rabbit
Antigen Affinity Chromatography
RUO
13178, 1633, 79127
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P27707, P43346, P48769
Dck
DCK Fusion Protein Ag12123
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales