missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYBRD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26735-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the cytochrome b(561) family that encodes an iron-regulated protein. It highly expressed in the duodenal brush border membrane. It has ferric reductase activity and is believed to play a physiological role in dietary iron absorption.Especificaciones
CYBRD1 | |
Polyclonal | |
Unconjugated | |
CYBRD1 | |
CYBRD1, cytochrome b reductase 1, DCYTB, Duodenal cytochrome b, Ferric chelate reductase 3, FRRS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79901 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.63 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q53TN4 | |
CYBRD1 | |
CYBRD1 Fusion Protein Ag24968 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |