missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL13/BCA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10927-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Chemotactic for B-lymphocytes but not for T-lymphocytes. monocytes and neutrophils. Does not induce calcium release in B-lymphocytes. Binds to BLR1/CXCR5. B lymphocyte chemoattractant. independently cloned and named Angie.Especificaciones
CXCL13/BCA1 | |
Polyclonal | |
Unconjugated | |
CXCL13 | |
ANGIE, ANGIE2, B cell attracting chemokine 1, B lymphocyte chemoattractant, BLC, BLR1L, C X C motif chemokine 13, CXC chemokine BLC, CXCL13, SCYB13, Small inducible cytokine B13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10563 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43927 | |
CXCL13 | |
CXCL13/BCA1 Fusion Protein Ag1358 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |