Learn More
CTC1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28268-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The CTC1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CTC1. This antibody recognizes human antigen. The CTC1 antibody has been validated for the following applications: WB,ELISA analysis.
Especificaciones
CTC1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
80169 | |
Liquid |
Western Blot, ELISA | |
267 μg/mL | |
Rabbit | |
Primary | |
Human | |
IgG |