missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTAGE6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17438-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
CTAGE6 | |
Polyclonal | |
Unconjugated | |
CTAGE6 | |
cTAGE family member 6, CTAGE family, member 6, CTAGE6, CTAGE6P, Putative protein cTAGE 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
340307 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86UF2 | |
CTAGE6 | |
CTAGE6 Fusion Protein Ag11511 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |