missing translation for 'onlineSavingsMsg'
Learn More

CSRP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10721-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16897283

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 21-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Cysteine and glycine-rich protein 3 (CSRP3) is a member of the CSRP family of genes encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in CRP3 is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Very strongly similar to rat MLP; contains zinc-finger LIM domains. Other genes in the family include CSRP1 and CSRP2.
Especificaciones

Especificaciones

CSRP3
Polyclonal
Unconjugated
CSRP3
Cardiac LIM protein, CLP, CMD1M, CMH12, CRP3, CSRP3, Cysteine rich protein 3, LIM domain protein, cardiac, LMO4, MLP, Muscle LIM protein
Rabbit
Antigen Affinity Chromatography
RUO
117505, 13009, 8048
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P50461, P50462, P50463
CSRP3
CSRP3 Fusion Protein Ag1136
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales