missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX18 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18071-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
COX18 encodes a cytochrome c oxidase (COX)-assembly protein. The S. cerevisiae Cox18 protein catalyzes the insertion of the Cox2 (MTCO2; MIM 516040) C-terminal tail into the mitochondrial inner membrane, an intermediate step in the assembly of complex IV of the mitochondrial respiratory chain. Required for the insertion of integral membrane proteins into the mitochondrial inner membrane. Essential for the activity and assembly of cytochrome c oxidase. Plays a central role in the translocation and export of the C-terminal part of the COX2 protein into the mitochondrial intermembrane space.Especificaciones
COX18 | |
Polyclonal | |
Unconjugated | |
COX18 | |
COX18, COX18HS, OXA1L2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231430, 285521 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N8Q8, Q8VC74 | |
COX18 | |
COX18 Fusion Protein Ag12709 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |