Learn More
STEMCELL Technologies CoV Protease Substrate-1 TF5
FRET-based peptide substrate for detection of protease activity of coronaviruses
715.54€ - 3049.69€
Especificaciones
Para utilizar con (aplicación) Enzyme Substrate; FRET
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17187631
Ver documentos Detalles de la promoción
STEMCELL Technologies
100-0505
100 Tests
715.54€
100 tests
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 24-01-2023
para ver el stock
Añadir a la cesta
17197631
Ver documentos Detalles de la promoción
STEMCELL Technologies
100-0506
1000 Tests
3049.69€
1000 tests
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 24-01-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • CoV Protease Substrate-1 TF5 allows for efficient detection of coronavirus (CoV) proteases
  • This fluorescence resonance energy transfer (FRET)-based peptide substrate is cleaved by 3C-like proteases (3CLpro), the CoV main protease involved in regulating the coronavirus replication complex
  • 3CLpro is also a potential target for therapeutic development
  • CoV Protease Substrate-1 TF5 has an excitation wavelength of 649 nm, an emission wavelength of 664 nm, and an extinction coefficient of 250,000 cm-1 M-1
  • It consists of a 14 amino acid sequence (KTSAVLQSGFRKME) that also contains the Tide Quencher™ 5 (TQ5; quencher) and Tide Fluor™ 5 (TF5; donor) fragments at the N- and C- termini, respectively
  • When the peptide substrate is intact, favorable energetic overlap of the TF5 excited state and the TQ5 absorption leads to efficient intramolecular fluorescence quenching of TF5 by the TQ5 fragment
  • Cleavage of the substrate by CoV proteases, however, dissociates the TF5 fragment from TQ5, restoring the fluorescence of the TF5 moiety that is directly correlated to the CoV protease activity
Especificaciones
Especificaciones
Enzyme Substrate; FRET