missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 32 Mouse, Clone: GJB1/1753, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 2705-MSM3-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Connexin 32 | |
Cytoplasmic and Cell Surface | |
GJB1/1753 | |
Unconjugated | |
XP_016884897 | |
Mouse | |
27-32kDa | |
Protein A/G Purified | |
RUO | |
2705 | |
IgG |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
GJB1 | |
Recombinant human GJB1 protein | |
MCF-7 Cells. Liver, Kidney, Stomach or Tonsil | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |