missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Collagenase Type 3 (C. histolyticum)
Used for Enzyme
Marca: Abnova™ P5278.50mg
Detalles adicionales : Peso : 0.00010kg
Descripción
#REF!Especificaciones
Enzyme Activity | |
Native protein purified from Clostridium histolyticum | |
Store at 4°C on dry atmosphere. For primary cell isolation, culture and tissue dissociation: Reconstituted in balanced salt solutions (such as Hank's, Earle's and others) at concentrations from 0.05% to 0.5% (w/v). For enzymetic assay: Reconstituted in 0.05 M TES buffer with 0.36 mM calcium chloride, pH 7.5 at a concentration of 1.0 mg/ml. | |
COLLAGENASE | |
≥100 units per mg dry weight. One Unit releases one micromole of L-leucine equivalents from collagen in 5 hours at 37°C, pH 7.5. | |
None | |
Lyophilized |
Collagenase Type 3 (C. histolyticum) | |
50 mg | |
RUO | |
Collagenase Assay; Mammary primary cell isolation. | |
Clostridium species | |
Native protein purified from Clostridium histolyticum |