missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11135-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The CCR4-NOT complex functions as general transcription regulation complex.Especificaciones
CNOT3 | |
Polyclonal | |
Unconjugated | |
CNOT3 | |
CCR4 associated factor 3, CNOT3, KIAA0691, LENG2, NOT3, NOT3H | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
232791, 308311, 4849 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75175, Q8K0V4 | |
CNOT3 | |
CNOT3 Fusion Protein Ag1587 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |