missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11092-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the CLK family of dual specificity protein kinases. CLK family members have been shown to interact with, and phosphorylate, serine- and arginine-rich (SR) proteins of the spliceosomal complex, which is a part of the regulatory mechanism that enables the SR proteins to control RNA splicing. Note that this gene is distinct from TELO2 gene (GeneID:9894), which shares CLK2 and hCLK2 symbol aliases in common with this gene, but encodes a protein that is involved in telomere length regulation.Especificaciones
CLK2 | |
Polyclonal | |
Unconjugated | |
CLK2 | |
CDC like kinase 2, CLK2, hCLK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1196, 12748, 365842 | |
-20°C | |
Liquid |
ELISA, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35491, P49760 | |
Clk2 | |
CLK2 Fusion Protein Ag1549 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |