missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST4 Mouse anti-Human, Clone: 1E1A4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66623-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an N-acetylglucosamine 6-O sulfotransferase. The encoded enzyme transfers sulfate from 3′phosphoadenosine 5′phospho-sulfate to the 6-hydroxyl group of N-acetylglucosamine on glycoproteins. This protein is localized to the Golgi and is involved in the modification of glycan structures on ligands of the lymphocyte homing receptor L-selectin. Alternate splicing in the 5′ UTR results in multiple transcript variants that encode the same protein.Especificaciones
CHST4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NCG5 | |
CHST4 | |
CHST4 Fusion Protein Ag3987 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
1E1A4 | |
Unconjugated | |
CHST4 | |
CHST4, GlcNAc6ST 2, Gn6st 2, GST 3, HEC GlcNAc6ST, LSST | |
Mouse | |
Protein G | |
RUO | |
10164 | |
-20°C | |
Liquid |