missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA5 Mouse anti-Human, Mouse, Porcine, Clone: 3G8C11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66363-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Nicotinic acetylcholine receptors (nAChRs), such as CHRNA5, are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are thought to be (hetero)pentamers composed of homologous subunits.Especificaciones
CHRNA5 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30532, Q2MKA5 | |
CHRNA5 | |
CHRNA5 Fusion Protein Ag4447 | |
150 μL | |
Primary | |
Human, Mouse, Porcine | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
3G8C11 | |
Unconjugated | |
CHRNA5 | |
CHRNA5, LNCR2, NACHRA5 | |
Mouse | |
Protein A | |
RUO | |
100156023, 110835, 1138 | |
-20°C | |
Liquid |