missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CHI3L2 (Human) Recombinant Protein
Human CHI3L2 full-length ORF ( AAH11460.1, 1 a.a. - 390 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001117-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MGATTMDQKSLWAGVVVLLLLQGGSAYKLVCYFTNWSQDRQEPGKFTPENIDPFLCSHLIYSFASIENNKVIIKDKSEVMLYQTINSLKTKNPKLKILLSIGGYLFGSKGFHPMVDSSTSRLEFINSIILFLRNHNFDGLDVSWIYPDQKENTHFTVLIHELAEAFQKDFTKSTKERLLLTVGVSAGRQMIDNSYQVEKLAKDLDFINLLSFDFHGSWEKPLITGHNSPLSKGWQDRGPSSYYNVEYAVGYWIHKGMPSEKVVMGIPTYGHSFTLASAETTVGAPASGPGAAGPITESSGFLAYYEICQFLKGAKITRLQDQQVPYAVKGNQWVGYDDVKSMETKVQFLKNLNLGGAMIWSIDMDDFTGKSCNQGPYPLVQAVKRSLGSL
Especificaciones
AAH11460.1 | |
chitinase 3-like 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHI3L2 | |
GST |
1117 | |
Wheat germ expression system | |
25 μg | |
YKL-39, YKL39 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |