missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CFHR1 (Human) Recombinant Protein
Human CFHR1 full-length ORF ( AAH16755.1, 19 a.a. - 330 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003078-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: EATFCDFPKINHGILYDEEKYKPFSQVPTGEVFYYSCEYNFVSPSKSFWTRITCTEEGWSPTPKCLRLCFFPFVENGHSESSGQTHLEGDTVQIICNTGYRLQNNENNISCVERGWSTPPKCRSTDTSCVNPPTVQNAHILSRQMSKYPSGERVRYECRSPYEMFGDEEVMCLNGNWTEPPQCKDSTGKCGPPPPIDNGDITSFPLSVYAPASSVEYQCQNLYQLEGNKRITCRNGQWSEPPKCLHPCVISREIMENYNIALRWTAKQKLYLRTGESAEFVCKRGYRLSSRSHTLRTTCWDGKLEYPTCAKR
Especificaciones
AAH16755.1 | |
complement factor H-related 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CFHR1 | |
GST |
3078 | |
Wheat germ expression system | |
10 μg | |
CFHL, CFHL1, CFHL1P, CFHR1P, FHR1, H36-1, H36-2, HFL1, HFL2, MGC104329 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |