Learn More
CENPC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14250-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.Especificaciones
CENPC | |
Polyclonal | |
Unconjugated | |
CENPC | |
CENP C, CENP C 1, CENPC, CENPC1, Centromere autoantigen C, centromere protein C 1, hcp 4, ICEN7, MIF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1060, 12617 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03188 | |
CENPC, Cenpc1 | |
CENPC Fusion Protein Ag5523 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |