missing translation for 'onlineSavingsMsg'
Learn More

CEACAM3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19496-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16819024

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 21-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The antibody is specific to isoform1 of CEACAM3.

This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Especificaciones

Especificaciones

CEACAM3
Polyclonal
Unconjugated
CEACAM3
Carcinoembryonic antigen CGM1, CD66D, CEACAM3, CGM1, W264, W282
Rabbit
Antigen Affinity Chromatography
RUO
1084
-20°C
Liquid
Immunohistochemistry (Paraffin), Flow Cytometry
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P40198
CEACAM3
Peptide
20 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales