missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19496-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody is specific to isoform1 of CEACAM3.
This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Especificaciones
CEACAM3 | |
Polyclonal | |
Unconjugated | |
CEACAM3 | |
Carcinoembryonic antigen CGM1, CD66D, CEACAM3, CGM1, W264, W282 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1084 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P40198 | |
CEACAM3 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |