missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDK5 (Human) Recombinant Protein
Human CDK5 full-length ORF ( AAH05115 , 1 a.a. - 292 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001020-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MQKYEKLEKIGEGTYGTVFKAKNRETHEIVALKRVRLDDDDEGVPSSALREICLLKELKHKNIVRLHDVLHSDKKLTLVFEFCDQDLKKYFDSCNGDLDPEIVKSFLFQLLKGLGFCHSRNVLHRDLKPQNLLINRNGELKLADFGLARAFGIPVRCYSAEVVTLWYRPPDVLFGAKLYSTSIDMWSAGCIFAELANAGRPLFPGNDVDDQLKRIFRLLGTPTEEQWPSMTKLPDYKPYPMYPATTSLVNVVPKLNATGRDLLQNLLKCNPVQRISAEEALQHPYFSDFCPP
Especificaciones
AAH05115 | |
cyclin-dependent kinase 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDK5 | |
GST |
1020 | |
Wheat germ expression system | |
10 μg | |
PSSALRE | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |