missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CDK11/cyclin C, GST Tag, His Tag Recombinant Protein
Human CDK11/cyclin C, GST Tag, His Tag Recombinant Protein
Marca: Invitrogen™ A33733
Detalles adicionales : Peso : 0.00900kg
Descripción
CDK11/cyclin C serine/threonine kinase is dimeric protein complex. CDK11 is one of the components of the Mediator co-activator complex. The Mediator complex is a multi-protein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. Cyclin C is a member of the cyclin family of proteins. The encoded protein interacts with cyclin-dependent kinase 8 and induces the phosphorylation of the carboxy-terminal domain of the large subunit of RNA polymerase II.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Especificaciones
CDK11: NP_055891.1, Cyclin C: NP_005181.2 | |
84.9/38.0 kDa | |
CDKs and CHKs | |
Human | |
GST Tag, His Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
1 mg | |
CDK11: 1 - 502, Cyclin C: 1 - 283 | |
Recombinant | |
Storage Buffer: 50 mM HEPES pH 7.5, 100 mM NaCl, 5 mM DTT, 20% glycerol | |
Baculovirus | |
Recombinant, Active, Catalytic |