missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDAN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17597-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that appears to play a role in nuclear envelope integrity, possibly related to microtubule attachments. Mutations in this gene cause congenital dyserythropoietic anemia type I, a disease resulting in morphological and functional abnormalities of erythropoiesis.Especificaciones
CDAN1 | |
Polyclonal | |
Unconjugated | |
CDAN1 | |
CDA I, CDA1, CDAI, CDAN1, codanin, Codanin 1, DLT, PRO1295 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
146059, 68968 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CC12, Q8IWY9 | |
CDAN1 | |
CDAN1 Fusion Protein Ag11589 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |