Learn More
CD90 (Thy-1) Mouse, anti-Human, PerCP-Cyanine5.5, Clone: 5E10, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60045PS.1
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17188761
  • en la oferta / 25 tests
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
Descripción
Descripción
  • The 5E10 antibody reacts with CD90 (Thy-1), a GPI-linked membrane glycoprotein that is N-glycosylated at two sites, giving rise to 25 - 37 kDa molecules
  • CD90 has roles in signal transduction, cell adhesion and migration, neurite outgrowth, T cell activation, tumor suppression, and inhibition of the proliferation and differentiation of hematopoietic stem cells
  • It is a known ligand of β2 and β3 integrins and upregulates synthesis of fibronectin, osteonectin and thrombospondin
  • CD90 is broadly expressed, being found on human thymocytes, neurons, some glial cells, fibroblasts, activated endothelial cells, some leukemia cell lines and a distinct subset (<1%) of CD3+CD4+ T cells in human peripheral blood
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Especificaciones
Especificaciones
CD90 (Thy-1)
Monoclonal
PerCP-Cyanine5.5
25 Tests
Baboon, Cynomolgus, Rhesus Monkey, Pig, Human
Hematopoietic Stem and Progenitor Cells, Mammary Cells, Mesenchymal Stem and Progenitor Cells
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
5E10
Mouse
Primary
IgG1 κ