Learn More
CD90 (Thy-1) Mouse, anti-Human, PerCP-Cyanine5.5, Clone: 5E10, STEMCELL Technologies
Mouse Monoclonal Antibody
319.20€ - 687.61€
Especificaciones
Antígeno CD90 (Thy-1)
Clon 5E10
Aplicaciones Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
Clasificación Monoclonal
Conjugado PerCP-Cyanine5.5
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17188761
Ver documentos Detalles de la promoción
Stemcell Technologies
60045PS.1
25 Tests
319.20€
25 tests
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 22-04-2024
para ver el stock
17178761
Ver documentos Detalles de la promoción
Stemcell Technologies
60045PS
100 Tests
687.61€
100 tests
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 22-04-2024
para ver el stock
Descripción
Descripción
  • The 5E10 antibody reacts with CD90 (Thy-1), a GPI-linked membrane glycoprotein that is N-glycosylated at two sites, giving rise to 25 - 37 kDa molecules
  • CD90 has roles in signal transduction, cell adhesion and migration, neurite outgrowth, T cell activation, tumor suppression, and inhibition of the proliferation and differentiation of hematopoietic stem cells
  • It is a known ligand of β2 and β3 integrins and upregulates synthesis of fibronectin, osteonectin and thrombospondin
  • CD90 is broadly expressed, being found on human thymocytes, neurons, some glial cells, fibroblasts, activated endothelial cells, some leukemia cell lines and a distinct subset (<1%) of CD3+CD4+ T cells in human peripheral blood
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Especificaciones
Especificaciones
CD90 (Thy-1)
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
PerCP-Cyanine5.5
Baboon, Cynomolgus, Rhesus Monkey, Pig, Human
Primary
5E10
Monoclonal
Mouse
IgG1 κ