missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD83 Mouse, anti-Human, Unconjugated, Clone: HB15e, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca: Stemcell Technologies 60107.1
Detalles adicionales : Peso : 0.00110kg
Descripción
- The HB15e antibody reacts with CD83, an ∼45 kDa type I transmembrane glycoprotein and member of the Siglec protein family, containing an extracellular V-type Ig-like domain and a C-terminal cytoplasmic tail
- A soluble form of CD83 has also been identified
- CD83 is expressed predominantly on circulating and tissue dendritic cells, including Langerhan cells and interdigitating reticulum cells
- It is expressed weakly on activated lymphocytes and may also be found on neutrophils, monocytes and macrophages
- Expression of CD83 is typically transient, except on dendritic cells, and correlates with upregulation of HLA class II antigen
- Its function, though ill-defined, is essential for the development and maintenance of the immune system and evidence suggests it may be involved in cell adhesion and signaling, antigen presentation, and lymphocyte activation
- The epitope on CD83 bound by the HB15e antibody reportedly overlaps with that of antibody 1E11
Especificaciones
CD83 | |
Monoclonal | |
Unconjugated | |
25 μg | |
Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
Dendritic Cells |
Immunoblot, Flow Cytometry, Immunohistochemistry | |
HB15e | |
Mouse | |
Primary | |
IgG1 κ |