missing translation for 'onlineSavingsMsg'
Learn More

CD83 Mouse, anti-Human, Unconjugated, Clone: HB15e, STEMCELL Technologies

Mouse Monoclonal Antibody

119.70€ - 291.27€

Especificaciones

Antígeno CD83
Clon HB15e
Aplicaciones Immunohistochemistry, Flow Cytometry, Immunoblot
Clasificación Monoclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17179051
Ver documentos Detalles de la promoción
Stemcell Technologies
60107
100 μg
291.27€
100 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 16-12-2022
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
17189051
Ver documentos Detalles de la promoción
Stemcell Technologies
60107.1
25 μg
119.70€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 16-12-2022
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

  • The HB15e antibody reacts with CD83, an ∼45 kDa type I transmembrane glycoprotein and member of the Siglec protein family, containing an extracellular V-type Ig-like domain and a C-terminal cytoplasmic tail
  • A soluble form of CD83 has also been identified
  • CD83 is expressed predominantly on circulating and tissue dendritic cells, including Langerhan cells and interdigitating reticulum cells
  • It is expressed weakly on activated lymphocytes and may also be found on neutrophils, monocytes and macrophages
  • Expression of CD83 is typically transient, except on dendritic cells, and correlates with upregulation of HLA class II antigen
  • Its function, though ill-defined, is essential for the development and maintenance of the immune system and evidence suggests it may be involved in cell adhesion and signaling, antigen presentation, and lymphocyte activation
  • The epitope on CD83 bound by the HB15e antibody reportedly overlaps with that of antibody 1E11
Especificaciones

Especificaciones

CD83
Immunohistochemistry, Flow Cytometry, Immunoblot
Unconjugated
Human, Chimpanzee, Rhesus Monkey, Cynomolgus, Baboon
Primary
HB15e
Monoclonal
Mouse
IgG1 κ
SDS
Product Certifications