missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: RPA-T8, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566452
Detalles adicionales : Peso : 0.01000kg
Descripción
CD8 Monoclonal antibody specifically detects CD8 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Especificaciones
CD8 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
RPA-T8 | |
Mouse | |
RUO | |
Human, Baboon, Cynomolgus, Rhesus Macaque | |
Purified |