missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD61 / Integrin beta-3 Mouse, Clone: Y2/51, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 3690-MSM2-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD61 / Integrin beta-3 | |
Cell Surface | |
Y2/51 | |
Unconjugated | |
NP_000203 | |
Mouse | |
105kDa ;90kDa | |
Protein A/G Purified | |
RUO | |
3690 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
ITGB3 | |
Phytohemagglutinin (PHA)-stimulated peripheral blood cells | |
U937, KG1a, HEL cells. Spleen | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |