missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17227-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter Syndrome.Especificaciones
CD5 | |
Polyclonal | |
Unconjugated | |
CD5 | |
CD5, CD5 molecule, LEU1, Lymphocyte antigen T1/Leu 1, T1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12507, 921 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06127, P13379 | |
CD5 | |
CD5 Fusion Protein Ag11065 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |