missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49b (Integrin α2) Rat, anti-Mouse, PE, Clone: DX5, STEMCELL Technologies
Rat Monoclonal Antibody
222.75€ - 569.70€
Specifications
| Antígeno | CD49b (Integrin α2) |
|---|---|
| Clon | DX5 |
| Aplicaciones | Immunohistochemistry, Flow Cytometry |
| Clasificación | Monoclonal |
| Conjugado | PE |
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
17188521
|
Stemcell Technologies
60020PE.1 |
50 μg |
222.75€
50 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17178521
|
Stemcell Technologies
60020PE |
200 μg |
569.70€
200 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
- The DX5 antibody reacts with murine CD49b (integrin α2), an ∼150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) to form the heterodimeric CD49b/CD29 complex known as VLA-2, a receptor for extracellular matrix proteins such as collagen, E-cadherin, fibronectin, and laminin
- CD49b is highly expressed by platelets and is found on a majority of NK cells, on NKT cells, and on a small subset of CD8+ T cells; the latter population increases substantially following viral infection
- CD49b is also expressed by several tissues, including intestine, kidney, mammary gland, and lung
- Binding of the DX5 antibody has not been observed to affect the function of the VLA-2 integrin
- DX5 binding is, however, blocked by the clone HMα2 antibody
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755)
Specifications
| CD49b (Integrin α2) | |
| Immunohistochemistry, Flow Cytometry | |
| PE | |
| Mouse | |
| Primary |
| DX5 | |
| Monoclonal | |
| Rat | |
| IgM κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title