missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49b (Integrin α2) Rat, anti-Mouse, PE, Clone: DX5, STEMCELL Technologies
Rat Monoclonal Antibody
Marca: Stemcell Technologies 60020PE
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The DX5 antibody reacts with murine CD49b (integrin α2), an ∼150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) to form the heterodimeric CD49b/CD29 complex known as VLA-2, a receptor for extracellular matrix proteins such as collagen, E-cadherin, fibronectin, and laminin
- CD49b is highly expressed by platelets and is found on a majority of NK cells, on NKT cells, and on a small subset of CD8+ T cells; the latter population increases substantially following viral infection
- CD49b is also expressed by several tissues, including intestine, kidney, mammary gland, and lung
- Binding of the DX5 antibody has not been observed to affect the function of the VLA-2 integrin
- DX5 binding is, however, blocked by the clone HMα2 antibody
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755)
Especificaciones
| CD49b (Integrin α2) | |
| Monoclonal | |
| PE | |
| 200 μg | |
| Mouse | |
| NK Cells |
| Immunohistochemistry, Flow Cytometry | |
| DX5 | |
| Rat | |
| Primary | |
| IgM κ |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido