missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.2 Mouse, FITC, Clone: 104, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio QAB43-F-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD45.2 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ptprc | |
Protein A/G Purified | |
RUO | |
19264 | |
IgG2a κ |
Flow Cytometry | |
104 | |
FITC | |
NP_001255215 | |
Mouse | |
500 μg | |
Primary | |
Store at 2-4°C. |