missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Rabbit, Clone: HPCA1/1806R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 947-RBM6-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD34 | |
Cell surface | |
HPCA1/1806R | |
Unconjugated | |
NP_001020280 | |
Rabbit | |
90-110kDa | |
Protein A/G Purified | |
RUO | |
947 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
CD34 | |
Recombinant full-length human HPCA1 protein | |
KG-1 cells, Tonsil, or Angiosarcoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |