missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Mouse anti-Human, Alexa Fluor 647, Clone: O323, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 567025
Dettagli aggiuntivi : Peso : 0.00720kg
Descrizione
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow cytometrySpecifica
CD27 | |
Monoclonal | |
Alexa Fluor 647 | |
CD27 antigen; TNFRSF7; S152; T14; Tp55 | |
Not Reported | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
O323 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |