Learn More
CD19 Mouse anti-Human, BB700, Clone: SJ25C1 (also known as SJ25-C1), BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566396
Detalles adicionales : Peso : 0.00900kg
Descripción
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Especificaciones
CD19 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
SJ25C1 (also known as SJ25-C1) | |
Mouse | |
RUO | |
Human | |
Purified |