missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Mouse, APC, Clone: PK136, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio QAB78-APC-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD161 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Klrb1c | |
Protein A/G Purified | |
RUO | |
17059 | |
IgG2a κ |
Flow Cytometry | |
PK136 | |
APC | |
NP_001153376 | |
Mouse | |
100 μg | |
Primary | |
Store at 2-4°C. |