missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Mouse anti-Human, BB700, Clone: MφP9 (also known as MφP-9), BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566465
Detalles adicionales : Peso : 0.00840kg
Descripción
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Especificaciones
CD14 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |
Flow Cytometry | |
MφP9 (also known as MφP-9) | |
Mouse | |
RUO | |
Human | |
Purified |