missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 / c-Kit Rat, APC-Cy7, Clone: ACK2, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB54-APC7-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD117 / c-Kit | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Kit | |
Protein A/G Purified | |
RUO | |
16590 | |
IgG2b κ |
Flow Cytometry | |
ACK2 | |
APC-Cyanine7 | |
NP_066922 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |