Learn More
CCDC70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 21558-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
CCDC70 | |
Polyclonal | |
Unconjugated | |
CCDC70 | |
CCDC70 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
498657, 67929, 83446 | |
-20°C | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6NSX1, Q9D9B0 | |
Ccdc70 | |
CCDC70 Fusion Protein Ag15600 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |