missing translation for 'onlineSavingsMsg'
Learn More

CASPR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 55417-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816885

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 21-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody is specific to CNTNAP1.

CNTNAP1 was initially identified as a 190-kDa protein associated with the contactin-PTPRZ1 complex. The 1, 384-amino acid protein, also designated p190 or CASPR for ′contactin-associated protein, ′ includes an extracellular domain with several putative protein-protein interaction domains, a putative transmembrane domain, and a 74-amino acid cytoplasmic domain. The architecture of its extracellular domain is similar to that of neurexins, and this protein may be the signaling subunit of contactin, enabling recruitment and activation of intracellular signaling pathways in neurons. The gene product was initially identified as a 190-kDa protein associated with the contactin-PTPRZ1 complex. The 1, 381-amino acid protein, also designated p190 or CASPR for ′contactin-associated protein, ′ includes an extracellular domain with several putative protein-protein interaction domains, a putative transmembrane domain, and a 74-amino acid cytoplasmic domain. Northern blot analysis showed that the gene is transcribed predomitly in brain as a transcript of 6.2 kb, with weak expression in several other tissues tested. The architecture of its extracellular domain is similar to that of neurexins, and this protein may be the signaling subunit of contactin, enabling recruitment and activation of intracellular signaling pathways in neurons.
Especificaciones

Especificaciones

CASPR
Polyclonal
Unconjugated
CNTNAP1
CASPR, Caspr1, CNTNAP, CNTNAP1, contactin associated protein 1, Neurexin 4, Neurexin IV, NRXN4, P190
Rabbit
Antigen Affinity Chromatography
RUO
53321, 84008, 8506
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54991, P78357, P97846
Cntnap1
Peptide
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales