missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASPR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | CASPR |
---|---|
Concentración | 0.4 mg/mL |
Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16816885
|
Proteintech
55417-1-AP-20UL |
20 μL |
140.00€
20 microlitros |
Fecha estimada de envío: 10-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16806885
|
Proteintech
55417-1-AP-150UL |
150 μL |
399.00€
150 microlitros |
Fecha estimada de envío: 10-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
This antibody is specific to CNTNAP1.
CNTNAP1 was initially identified as a 190-kDa protein associated with the contactin-PTPRZ1 complex. The 1, 384-amino acid protein, also designated p190 or CASPR for ′contactin-associated protein, ′ includes an extracellular domain with several putative protein-protein interaction domains, a putative transmembrane domain, and a 74-amino acid cytoplasmic domain. The architecture of its extracellular domain is similar to that of neurexins, and this protein may be the signaling subunit of contactin, enabling recruitment and activation of intracellular signaling pathways in neurons. The gene product was initially identified as a 190-kDa protein associated with the contactin-PTPRZ1 complex. The 1, 381-amino acid protein, also designated p190 or CASPR for ′contactin-associated protein, ′ includes an extracellular domain with several putative protein-protein interaction domains, a putative transmembrane domain, and a 74-amino acid cytoplasmic domain. Northern blot analysis showed that the gene is transcribed predomitly in brain as a transcript of 6.2 kb, with weak expression in several other tissues tested. The architecture of its extracellular domain is similar to that of neurexins, and this protein may be the signaling subunit of contactin, enabling recruitment and activation of intracellular signaling pathways in neurons.Especificaciones
CASPR | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O54991, P78357, P97846 | |
53321, 84008, 8506 | |
Peptide | |
Primary | |
-20°C | |
CNTNAP1 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CASPR, Caspr1, CNTNAP, CNTNAP1, contactin associated protein 1, Neurexin 4, Neurexin IV, NRXN4, P190 | |
Cntnap1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |