Learn More
Gasa Carpenter™ 20, malla 100 tejida a partir de alambre de 0,1 mm de diámetro, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar 046415.FL
Detalles adicionales : Peso : 0.01000kg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Carpenter 20™ gauze | |
75 x 75 mm | |
Odorless |
100 mesh woven from 0.1mm (0.004 in.) dia. wire | |
100 Mesh | |
Tela de alambre |