missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Canine GRO alpha/CXCL1 ELISA Kit

Canine GRO alpha/CXCL1 quantitates canine GRO alpha/CXCL1 in serum, plasma, supernatant.

Marca:  Invitrogen™ EC6RBX10

3041.80 EUR hasta el 2023-03-27
Use el código promocional "20585" para aplicar la oferta.



Detalles adicionales : Peso : 2.50000kg

 Ver más versiones de este producto

Código de producto. 17874040

  • 4540.00€ / paquete de 10

Fecha estimada de envío: 24-04-2023
para ver el stock
Añadir a la cesta

Descripción

Descripción

Canine GRO alpha/CXCL1 quantitates canine GRO alpha/CXCL1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant canine GRO alpha/CXCL1.

CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
Especificaciones

Especificaciones

alpha-chemokine; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; Cxcl1; CXCL-1; cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; FSP; gro; GRO a; GRO a; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROa; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; Mgsa; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51
5.4 pg/mL
ELISA
Colorimetric Microplate Reader
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
5.4 to 350 pg/mL
Biotin
Supernatant
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
CXCL1 (GRO alpha)
4 hr 45 min
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales