missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin-1 Rabbit, Clone: CNN1/1408R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 1264-RBM4-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Calponin-1 | |
Cytoplasmic | |
CNN1/1408R | |
Unconjugated | |
XP_005259798 | |
Rabbit | |
34kDa | |
Protein A/G Purified | |
RUO | |
1264 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
CNN1 | |
Recombinant human full-length CNN1 protein | |
Myoepithelial cells in breast ducts | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |