missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-17 Mouse anti-Human, Porcine, Clone: 3B10A9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60351-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Cadherins are calcium dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types.Especificaciones
Cadherin-17 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12864 | |
CDH17 | |
Cadherin-17 Fusion Protein Ag19488 | |
20 μL | |
Primary | |
Porcine, Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
3B10A9 | |
Unconjugated | |
CDH17 | |
Cadherin-17, CDH16, CDH17, HPT 1, HPT1, LI cadherin, Liver intestine cadherin | |
Mouse | |
Protein G | |
RUO | |
100157834, 1015 | |
-20°C | |
Liquid |