missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf61 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25630-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
C9orf61, also named as FAM189A2 and X123, is predomitly expressed in muscle. It has three isoforms with MW 45-50 kDa, 32-35 kDa and 27 kDa.Especificaciones
C9orf61 | |
Polyclonal | |
Unconjugated | |
FAM189A2 | |
C9orf61, FAM189A2, Protein FAM189A2, Protein X123, RP11 548B3.1, X123 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
381217, 9413 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15884, Q4FZH1 | |
Fam189a2 | |
C9orf61 Fusion Protein Ag22242 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |