missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1R Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17346-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody recognizes full-length C1r chain and the cleaved B chain.
C1r B chain is a serine protease that combines with C1q and C1s to form C1, the first component of the classical pathway of the complement system.Especificaciones
C1R | |
Polyclonal | |
Unconjugated | |
C1R | |
C1R, Complement C1r subcomponent | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
715 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P00736 | |
C1R | |
C1R Fusion Protein Ag11390 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |