missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf36 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25650-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
IZUMO4 is a protein coding gene.Especificaciones
C19orf36 | |
Polyclonal | |
Unconjugated | |
IZUMO4 | |
C19orf36, IMAGE:4215339, IZUMO4, Sperm 22 kDa protein c113 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113177, 71564 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D3Z690, Q1ZYL8 | |
Izumo4 | |
C19orf36 Fusion Protein Ag20434 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |