missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTNL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25110-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
BTNL2 (butyrophilin-like protein 2), also known as BTL-II, is a 455 amino acid single-pass type II membrane protein that acts as a negative regulator of T cell proliferation. Expressed in thymus, brain, kidney, heart, liver, small intestine, testis, ovary, pancreas and leukocytes, BTNL2 exists as at least six alternatively spliced isoforms, which are encoded by a gene located on human chromosome 6. Expression of BTNL2 isoform 3 is associated with susceptibility to sarcoidosis type 2 (SS2), an immune disorder that causes chronic inflammatory granulomatous lesions.Especificaciones
BTNL2 | |
Polyclonal | |
Unconjugated | |
BTNL2 | |
BTL II, BTNL2, Butyrophilin like protein 2, HSBLMHC1, SS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
547431, 56244 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70355, Q9UIR0 | |
Btnl2 | |
BTNL2 Fusion Protein Ag18803 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |